Learn More
Azzota Corp Standard Benchtop Turbidity Meter With Total Suspended Solids Measurement Mode, 2 To 7 Points Calibration, Selectable 4 Turbidity Units.
Catalog No. 501613019
Encompass_Preferred
Standard benchtop turbidity meter with Total Suspended Solids measurement mode, 2 to 7 points calibration, selectable 4 turbidity units.
Catalog No. 50-161-3019 Supplier Azzota Corp Supplier No. TMB200
Standard benchtop turbidity meter with Total Suspended Solids measurement mode, 2 to 7 points calibration, selectable 4 turbidity units.