Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-BRD9500 | 1630760-76-7 | 99.5% | C15H18FN3O2 | 5 MG
SDP

Catalog No. 5000222497
Encompass_Preferred

(S)-BRD9500 is the S-enantiomer of BRD9500 provided for research use as an experimental control in studies of phosphodiesterase 3 (PDE3) inhibition. It is supplied in small quantities for in vitro and in vivo assays, with storage and purity specifications suitable for laboratory workflows.

  • S enantiomer used as an experimental control in PDE3 inhibition studies.
  • High reported purity (~99.5% by HPLC).
  • Molecular formula C15H18FN3O2 and molecular weight 291.32 g·mol-1.
  • Available in small research quantities (1 mg, 5 mg, 10 mg).
  • Store powder at -20°C long term or 4°C short term; store solutions at -80°C long term.

Catalog No. 50-002-22497 Supplier Medchemexpress LLC Supplier No. HY136350B5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S)-BRD9500 is the S-enantiomer of BRD9500 provided for research use as an experimental control in studies of phosphodiesterase 3 (PDE3) inhibition. It is supplied in small quantities for in vitro and in vivo assays, with storage and purity specifications suitable for laboratory workflows.

  • S enantiomer used as an experimental control in PDE3 inhibition studies.
  • High reported purity (~99.5% by HPLC).
  • Molecular formula C15H18FN3O2 and molecular weight 291.32 g·mol-1.
  • Available in small research quantities (1 mg, 5 mg, 10 mg).
  • Store powder at -20°C long term or 4°C short term; store solutions at -80°C long term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.