Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-bromoenol lactone | 478288-94-7 | 99.1% | 1 MG
SDP

Catalog No. 5000232796
Encompass_Preferred

(S)-Bromoenol lactone is a chiral, mechanism-based irreversible inhibitor of calcium-independent phospholipase A2β (iPLA2β) used in biochemical and cellular research to investigate arachidonate release pathways. The compound is supplied at high purity as a solution and is typically stored at -20°C for long-term stability.

  • Irreversible inhibitor of calcium-independent phospholipase A2β (iPLA2β)
  • Reported activity: IC50 ≈ 2 μM for vasopressin-induced arachidonate release in A10 cells
  • Molecular weight 317.18 g/mol; formula C16H13BrO2
  • Purity 99.1%
  • Storage: solution at -20°C; stable for about 2 years

Catalog No. 50-002-32796 Supplier Medchemexpress LLC Supplier No. HY1209861MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S)-Bromoenol lactone is a chiral, mechanism-based irreversible inhibitor of calcium-independent phospholipase A2β (iPLA2β) used in biochemical and cellular research to investigate arachidonate release pathways. The compound is supplied at high purity as a solution and is typically stored at -20°C for long-term stability.

  • Irreversible inhibitor of calcium-independent phospholipase A2β (iPLA2β)
  • Reported activity: IC50 ≈ 2 μM for vasopressin-induced arachidonate release in A10 cells
  • Molecular weight 317.18 g/mol; formula C16H13BrO2
  • Purity 99.1%
  • Storage: solution at -20°C; stable for about 2 years

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.