Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-Butoxycarbonylamino-cyclopropyl-acetic acid | 155976-13-9 | 98.0% | C10H17NO4 | 10 G
SDP

Catalog No. 5000434201
Encompass_Preferred

(S)-Butoxycarbonylamino-cyclopropyl-acetic acid is a Glycine derivative. It is for research use only and not sold to patients. Amino acids and amino acid derivatives, such as this compound, are commercially used as ergogenic supplements. They can influence the secretion of anabolic hormones, supply fuel during exercise, improve mental performance under stress, and prevent exercise-induced muscle damage.

  • Molecular weight: 215.25
  • Formula: C10H17NO4
  • CAS No.: 155976-13-9
  • Appearance: Solid
  • Color: White to off-white
  • Purity: 98.0%
  • Shipping: Room temperature in continental US
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Solubility (in vitro) in DMSO: 100 mg/mL (464.58 mM)

Catalog No. 50-004-34201 Supplier Medchemexpress LLC Supplier No. HY6026410G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S)-Butoxycarbonylamino-cyclopropyl-acetic acid is a Glycine derivative. It is for research use only and not sold to patients. Amino acids and amino acid derivatives, such as this compound, are commercially used as ergogenic supplements. They can influence the secretion of anabolic hormones, supply fuel during exercise, improve mental performance under stress, and prevent exercise-induced muscle damage.

  • Molecular weight: 215.25
  • Formula: C10H17NO4
  • CAS No.: 155976-13-9
  • Appearance: Solid
  • Color: White to off-white
  • Purity: 98.0%
  • Shipping: Room temperature in continental US
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Solubility (in vitro) in DMSO: 100 mg/mL (464.58 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.