Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-Butoxycarbonylamino-cyclopropyl-acetic acid | 155976-13-9 | 98.0% | C10H17NO4 | 5 G
SDP

Catalog No. 5000433988
Encompass_Preferred

(S)-Butoxycarbonylamino-cyclopropyl-acetic acid is a Glycine derivative that has been studied for its use as an ergogenic supplement. It is recognized for its potential to influence anabolic hormone secretion, supply fuel during exercise, enhance mental performance under stress, and prevent exercise-induced muscle damage.

  • High purity: 98.0%
  • Molecular weight: 215.25
  • Chemical formula: C10H17NO4
  • White to off-white solid appearance
  • Soluble in DMSO (100 mg/mL)
  • Stable in powder form at -20°C for 3 years, 4°C for 2 years
  • Stable in solvent at -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-33988 Supplier Medchemexpress LLC Supplier No. HY602645G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S)-Butoxycarbonylamino-cyclopropyl-acetic acid is a Glycine derivative that has been studied for its use as an ergogenic supplement. It is recognized for its potential to influence anabolic hormone secretion, supply fuel during exercise, enhance mental performance under stress, and prevent exercise-induced muscle damage.

  • High purity: 98.0%
  • Molecular weight: 215.25
  • Chemical formula: C10H17NO4
  • White to off-white solid appearance
  • Soluble in DMSO (100 mg/mL)
  • Stable in powder form at -20°C for 3 years, 4°C for 2 years
  • Stable in solvent at -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.