Learn More
AdipoGen S-Butyrylthiocholine chloride
SDP
Catalog No. 502561696
Encompass_Preferred
Chemical. CAS 22026-63-7. Formula C9H20ClNOS. MW 225.78. Synthetic. Substrate to assay butyrylcholinesterase BuChE inhibitory activity. Also used as a building block for synthesis.
Catalog No. 50-256-1696 Supplier AdipoGen Supplier No. CDXB0041G001
Chemical. CAS 22026-63-7. Formula C9H20ClNOS. MW 225.78. Synthetic. Substrate to assay butyrylcholinesterase BuChE inhibitory activity. Also used as a building block for synthesis.