Learn More
Bellco Glass, Inc. Screw Cap 3 Baffles Fernbach, 1800mL With Blue PP Cap
SDP
Catalog No. 501945553
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Designed for culturing organisms that require a large surface area to volume ratio. Flask is supplied with a GL45 blue polypropylene linerless screw cap and pouring ring.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 1800mlOD X OAH (MM):200 X 157Case Quantity: 1
Catalog No. 50-194-5553 Supplier Bellco Glass, Inc. Supplier No. 255201800
Manufactured from Type 1 Class A Borosilicate Glass. Designed for culturing organisms that require a large surface area to volume ratio. Flask is supplied with a GL45 blue polypropylene linerless screw cap and pouring ring.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 1800mlOD X OAH (MM):200 X 157Case Quantity: 1