Learn More
Bellco Glass, Inc. Screw Cap 3 Bottom Baffles Fernbach,1800mL With Membrane Cap
SDP
Catalog No. 501945554
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Designed for culturing organisms that require a large surface area to volume ratio. Flask is supplied with a GL45 blue polypropylene linerless screw cap and pouring ring.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 1800mlOD X OAH (MM):200 X 157Case Quantity: 1
Catalog No. 50-194-5554 Supplier Bellco Glass, Inc. Supplier No. 255201803
Manufactured from Type 1 Class A Borosilicate Glass. Designed for culturing organisms that require a large surface area to volume ratio. Flask is supplied with a GL45 blue polypropylene linerless screw cap and pouring ring.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 1800mlOD X OAH (MM):200 X 157Case Quantity: 1