Learn More
Bellco Glass, Inc. Screw Cap 3Baf Nephelo Flask,500mL
SDP
Catalog No. 501945584
Encompass_Preferred
Ideal for growing, storing and mixing of aerobic and anaerobic cultures. Has a threaded neck and sidearm with clean-out port. 3 baffles increase oxygenation of cultures. Supplied with 38mm and 18mm rubber lined black phenolic cap.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 500mlIncludes: 38mm & 18mm Rubber Lined Black Phenolic CapSidearm Dimensions: 19x130mmCase Quantity: 4
Catalog No. 50-194-5584 Supplier Bellco Glass, Inc. Supplier No. 258114135
Ideal for growing, storing and mixing of aerobic and anaerobic cultures. Has a threaded neck and sidearm with clean-out port. 3 baffles increase oxygenation of cultures. Supplied with 38mm and 18mm rubber lined black phenolic cap.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 500mlIncludes: 38mm & 18mm Rubber Lined Black Phenolic CapSidearm Dimensions: 19x130mmCase Quantity: 4