Learn More
Bellco Glass, Inc. Screw Cap 45mm Erlenmeyer Flask, 1000mL Graduated
SDP
Catalog No. 501945485
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Ideal for all cell culture and media storage purposes. Provides flexible secure seal without a liner. Supplied with gl45 liner-less screw cap and pouring rings. Graduated to approximate volumes.. MADE IN THE USA Size: 1L Dimensions: 128x200mm
Catalog No. 50-194-5485 Supplier Bellco Glass, Inc. Supplier No. 251501000
Manufactured from type 1 class a Borosilicate glass. Ideal for all cell culture and media storage purposes. Provides flexible secure seal without a liner. Supplied with gl45 liner-less screw cap and pouring rings. Graduated to approximate volumes.. MADE IN THE USA Size: 1L Dimensions: 128x200mm