Learn More
Bellco Glass, Inc. Screw Cap 45mm Erlenmeyer Flask, 4000mL
SDP
Supplier:  Bellco Glass, Inc. 251504000
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Ideal for all cell culture and media storage purposes. Provides flexible secure seal without a liner. Supplied with gl45 liner-less screw cap and pouring rings. Graduated to approximate volumes.. MADE IN THE USA Size: 4L Dimensions: 205x325mm
Catalog No. 50-194-5487