Learn More
Bellco Glass, Inc. Screw Cap 45mm Erlenmeyer Flask, 6000mL
SDP
Catalog No. 501945488
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Ideal for all cell culture and media storage purposes. Provides flexible secure seal without a liner. Supplied with gl45 liner-less screw cap and pouring rings. Graduated to approximate volumes. Size: 6L Dimensions: 235x370mm
Catalog No. 50-194-5488 Supplier Bellco Glass, Inc. Supplier No. 251506000
Manufactured from type 1 class a Borosilicate glass. Ideal for all cell culture and media storage purposes. Provides flexible secure seal without a liner. Supplied with gl45 liner-less screw cap and pouring rings. Graduated to approximate volumes. Size: 6L Dimensions: 235x370mm