Learn More
Bellco Glass, Inc. Screw Cap Cult Tube Complete, 13x100mm With Bellco logo
SDP
Catalog No. 501945295
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Includes long skirted black phenolic screw cap with white rubber liner. MADE IN THE USA. Screw cap included Size: 13 x 100mm Replacement cap: 2029-00013
Catalog No. 50-194-5295 Supplier Bellco Glass, Inc. Supplier No. 201213100
Manufactured from type 1 class a Borosilicate glass. Includes long skirted black phenolic screw cap with white rubber liner. MADE IN THE USA. Screw cap included Size: 13 x 100mm Replacement cap: 2029-00013