Learn More
Bellco Glass, Inc. Screw Cap Cult Tube Complete, 16X150mm With Bellco logo
SDP
Catalog No. 501945297
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Includes long skirted black phenolic screw cap with white rubber liner. MADE IN THE USA. Screw cap included Size: 16 x 150mm Replacement cap: 2029-00016
Catalog No. 50-194-5297 Supplier Bellco Glass, Inc. Supplier No. 201216150
Manufactured from type 1 class a Borosilicate glass. Includes long skirted black phenolic screw cap with white rubber liner. MADE IN THE USA. Screw cap included Size: 16 x 150mm Replacement cap: 2029-00016