Learn More
Bellco Glass, Inc. Screw Cap Cult Tube Only,13X100mm Bellco Logo
SDP
Catalog No. 501945292
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Includes long skirted black phenolic screw cap with white rubber liner. MADE IN THE USA. Screw cap included Size: 13 x 100mm Replacement cap: 2029-00013
Catalog No. 50-194-5292 Supplier Bellco Glass, Inc. Supplier No. 201201310
Add to Cart
Add to Cart
Manufactured from type 1 class a Borosilicate glass. Includes long skirted black phenolic screw cap with white rubber liner. MADE IN THE USA. Screw cap included Size: 13 x 100mm Replacement cap: 2029-00013