Learn More
Bellco Glass, Inc. Screw Cap Culture Tube Only,38x200mm
SDP
Catalog No. 501945425
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass, with a round bottom. Tube only. 38x200mm. MADE IN THE USA
Catalog No. 50-194-5425 Supplier Bellco Glass, Inc. Supplier No. 211203820
Manufactured from type 1 class a Borosilicate glass, with a round bottom. Tube only. 38x200mm. MADE IN THE USA