Learn More
Bellco Glass, Inc. Screw Cap Culture Tube Complete, 20x150mm Bulk-Pack
SDP
Catalog No. 501945298
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Includes long skirted black phenolic screw cap with white rubber liner. MADE IN THE USA. Screw cap included Size: 20 x 150mm Replacement cap: 2029-00018
Catalog No. 50-194-5298 Supplier Bellco Glass, Inc. Supplier No. 201220000
Manufactured from type 1 class a Borosilicate glass. Includes long skirted black phenolic screw cap with white rubber liner. MADE IN THE USA. Screw cap included Size: 20 x 150mm Replacement cap: 2029-00018