Learn More
Bellco Glass, Inc. Screw Cap Fernbach W/O Baffles2800mL With Membrane Cap
SDP
Catalog No. 501945559
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Designed for culturing organisms that require a large surface area to volume ratio. Flask is supplied with a GL45 blue polypropylene linerless screw cap and pouring ring. WITHOUT BAFFLES. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 2800mlOD X OAH (MM):210 X 225Case Quantity: 1
Catalog No. 50-194-5559 Supplier Bellco Glass, Inc. Supplier No. 255228000
Manufactured from Type 1 Class A Borosilicate Glass. Designed for culturing organisms that require a large surface area to volume ratio. Flask is supplied with a GL45 blue polypropylene linerless screw cap and pouring ring. WITHOUT BAFFLES. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 2800mlOD X OAH (MM):210 X 225Case Quantity: 1