Learn More
Chemglass Life Sciences Flask, Fernbach, 2800mL, WITHOUT Baffles, GL-45 Thread, Vented Cap, Approx OD x Height (mm): 210 x 230
SDP
Catalog No. 501214780
Encompass_Preferred
Flask, Fernbach, 2800mL, WITHOUT Baffles, GL-45 Thread, Vented Cap, Approx OD x Height (mm): 210 x 230. Flasks are designed for culturing organisms that require a large surface area to volume ratio.
Catalog No. 50-121-4780 Supplier Chemglass Life Sciences Supplier No. CLS202030
Flask, Fernbach, 2800mL, WITHOUT Baffles, GL-45 Thread, Vented Cap, Approx OD x Height (mm): 210 x 230. Flasks are designed for culturing organisms that require a large surface area to volume ratio.