Learn More
Bellco Glass, Inc. Screw Cap Fernbach Flask w/6 Baff,2.8L
SDP
Catalog No. 501945557
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Designed for culturing organisms that require a large surface area to volume ratio. Flask is supplied with a GL45 blue polypropylene linerless screw cap and pouring ring.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 2800mlOD X OAH (MM):210 X 225Case Quantity: 1
Catalog No. 50-194-5557 Supplier Bellco Glass, Inc. Supplier No. 255202833
Manufactured from Type 1 Class A Borosilicate Glass. Designed for culturing organisms that require a large surface area to volume ratio. Flask is supplied with a GL45 blue polypropylene linerless screw cap and pouring ring.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 2800mlOD X OAH (MM):210 X 225Case Quantity: 1