Learn More
Enterprise Technology Solutions S.C. JOHNSON & SON, INC Air Freshener Refill,f/AutoSpray,CleanLinen,6.2oz,Clear
SDP
Supplier:  Enterprise Technology Solutions SJN375364
Encompass_Preferred
Add a burst of boldness and make your space your own with this automatic spray refill. Long-lasting design freshens rooms for up to 60 days when set at the 36-minute setting for cost-efficient performance. Refill is perfect for use in busy bathrooms, kitchens, lobbies and more. Automatic spray refill is designed for use with Glad automatic sprayers (sold separately).
Catalog No. 50-007-90671