Learn More
Enterprise Technology Solutions S.C. JOHNSON & SON, INC Dispenser, Manual, 1 Liter, 5-1/10"x4-3/5"x9-1/5", White
SDP
Catalog No. 5000789878
Encompass_Preferred
Proline Curve Manual Dispenser for washrooms features durable, plastic components and is rigorously tested for high-usage environments to provide assurance of quality and long-term cost-efficiency. Dispenser push button is exclusively protected by BioCote, a silver-based antimicrobial agent that is incorporated into the plastic components at the time of manufacture and inhibits the growth of a broad spectrum of bacteria and molds. BioCote will not wear out or rub off and is effective for the lifetime of the dispenser. Fixed product dose of soaps and shower products delivers the exact amount required to control usage and waste. Ultrasonically sealed cartridge and pump mechanisms prevent the ingress of air into the cartridge during use to help prevent product contamination and ensure fresh product is dispensed every time. Manual dispenser is designed for all S.C. Johnson Professional 1 liter manual cartridges (sold separately).
Catalog No. 50-007-89878 Supplier Enterprise Technology Solutions Supplier No. SJNWHB1LDS
Proline Curve Manual Dispenser for washrooms features durable, plastic components and is rigorously tested for high-usage environments to provide assurance of quality and long-term cost-efficiency. Dispenser push button is exclusively protected by BioCote, a silver-based antimicrobial agent that is incorporated into the plastic components at the time of manufacture and inhibits the growth of a broad spectrum of bacteria and molds. BioCote will not wear out or rub off and is effective for the lifetime of the dispenser. Fixed product dose of soaps and shower products delivers the exact amount required to control usage and waste. Ultrasonically sealed cartridge and pump mechanisms prevent the ingress of air into the cartridge during use to help prevent product contamination and ensure fresh product is dispensed every time. Manual dispenser is designed for all S.C. Johnson Professional 1 liter manual cartridges (sold separately).