Learn More
Enterprise Technology Solutions S.C. JOHNSON & SON, INC Dispenser,Manual,f/Moisturizing Cream,1 L Cap,White/Green
SDP
Catalog No. 5000830144
Encompass_Preferred
Use this Moisturizing Cream Dispenser in the industrial, commercial, healthcare and foodservice/processing facilities. It holds 1-liter SC Johnson Professional Stokolan and SBS 40 restore product cartridges. Large button is easy to push by hand or elbow so it is suitable in hygiene-sensitive areas. It dispenses a controlled dose to reduce waste. Color-coded push button is green for easy identification to encourage correct product usage and compliance. Dispenser button is exclusively protected with BioCote, a silver-based antimicrobial agent that is incorporated into plastic components at the time of manufacture and inhibits the growth of a broad spectrum of bacteria and molds. BioCote will not wear out or rub off and is effective for the lifetime of the dispenser button. Sight glass indicates the level of moisturizing in the cartridge that helps identify when product needs replacing. Unique locking mechanism helps prevent unauthorized access, pilferage and product contamination.
Catalog No. 50-008-30144 Supplier Enterprise Technology Solutions Supplier No. SJNRES1LDS
Use this Moisturizing Cream Dispenser in the industrial, commercial, healthcare and foodservice/processing facilities. It holds 1-liter SC Johnson Professional Stokolan and SBS 40 restore product cartridges. Large button is easy to push by hand or elbow so it is suitable in hygiene-sensitive areas. It dispenses a controlled dose to reduce waste. Color-coded push button is green for easy identification to encourage correct product usage and compliance. Dispenser button is exclusively protected with BioCote, a silver-based antimicrobial agent that is incorporated into plastic components at the time of manufacture and inhibits the growth of a broad spectrum of bacteria and molds. BioCote will not wear out or rub off and is effective for the lifetime of the dispenser button. Sight glass indicates the level of moisturizing in the cartridge that helps identify when product needs replacing. Unique locking mechanism helps prevent unauthorized access, pilferage and product contamination.