Learn More
Enterprise Technology Solutions S.C. JOHNSON & SON, INC Dispenser,Manual,Foam,Proline Curved,2L,BK
SDP
Catalog No. 5000789875
Encompass_Preferred
Use this Proline Curve Manual Dispenser in any office or public facility environment. It delivers the exact amount required, controlling usage and reducing waste. The 2-liter capacity holds foam SC Johnson Professional refill product cartridges. Perfect for hygiene-sensitive areas, large button is easy to push by hand or elbow for a controlled dose to reduce waste. Dispenser button is exclusively protected with BioCote, a silver-based antimicrobial agent that is incorporated into plastic components and inhibits the growth of a broad spectrum of bacteria and molds. BioCote will not wear out or rub off and is effective for the lifetime of the dispenser button.
Catalog No. 50-007-89875 Supplier Enterprise Technology Solutions Supplier No. SJNBLK2LDP
Use this Proline Curve Manual Dispenser in any office or public facility environment. It delivers the exact amount required, controlling usage and reducing waste. The 2-liter capacity holds foam SC Johnson Professional refill product cartridges. Perfect for hygiene-sensitive areas, large button is easy to push by hand or elbow for a controlled dose to reduce waste. Dispenser button is exclusively protected with BioCote, a silver-based antimicrobial agent that is incorporated into plastic components and inhibits the growth of a broad spectrum of bacteria and molds. BioCote will not wear out or rub off and is effective for the lifetime of the dispenser button.