Learn More
Enterprise Technology Solutions S.C. JOHNSON & SON, INC Dispenser, Touch Free, Foam Soap/Sanitizer, 2L, Black
SDP
Catalog No. 5000829065
Encompass_Preferred
TouchFREE Ultra Dispenser is a power-efficient, touch-free dispensing system for use with SCJ Professional foaming, hand hygiene products. Touch-free dispensing encourages use and minimizes any risk of cross-contamination. Infra-red sensing technology ensures reliable operation to prevent misfiring. Large capacity is made for 1.2 liters of SC Johnson Professional foam soap or non-alcohol sanitizer (approximately 1,700 applications) and 1 liter of SC Johnson Professional foam alcohol hand sanitizer (1,400 applications). Exceptional long battery life means a set of four D batteries will last up to 5 years of operation in typical usage conditions. Use this locking, compact dispenser on walls in washrooms, corridors and hallways or on SC Johnson Professional's freestanding stand (not included). Dispenser can be converted to open with a release button (included) for easy access.
Catalog No. 50-008-29065 Supplier Enterprise Technology Solutions Supplier No. SJNTF2BLK
TouchFREE Ultra Dispenser is a power-efficient, touch-free dispensing system for use with SCJ Professional foaming, hand hygiene products. Touch-free dispensing encourages use and minimizes any risk of cross-contamination. Infra-red sensing technology ensures reliable operation to prevent misfiring. Large capacity is made for 1.2 liters of SC Johnson Professional foam soap or non-alcohol sanitizer (approximately 1,700 applications) and 1 liter of SC Johnson Professional foam alcohol hand sanitizer (1,400 applications). Exceptional long battery life means a set of four D batteries will last up to 5 years of operation in typical usage conditions. Use this locking, compact dispenser on walls in washrooms, corridors and hallways or on SC Johnson Professional's freestanding stand (not included). Dispenser can be converted to open with a release button (included) for easy access.