Learn More
Enterprise Technology Solutions S.C. JOHNSON & SON, INC Dispensing System,f/Floor Cleaner,w/Cartridge,Concentrate.RD
SDP
Catalog No. 5000789874
Encompass_Preferred
TruFill Dispensing System easily replaces wall-mounted dispensing systems. Heavy-Duty Neutral Floor Cleaner comes concentrated in a portable, no-spill cartridge. Specially designed, integrated metering system in each cartridge provides worry-free dilution and delivery. One cartridge delivers up to 271 in-use gallons. Use cartridges and dispenser along-side the TruFill hose (not included).
Catalog No. 50-007-89874 Supplier Enterprise Technology Solutions Supplier No. SJN684501
TruFill Dispensing System easily replaces wall-mounted dispensing systems. Heavy-Duty Neutral Floor Cleaner comes concentrated in a portable, no-spill cartridge. Specially designed, integrated metering system in each cartridge provides worry-free dilution and delivery. One cartridge delivers up to 271 in-use gallons. Use cartridges and dispenser along-side the TruFill hose (not included).