Learn More
Enterprise Technology Solutions S.C. JOHNSON & SON, INC Floor Cleaner,Concentrate,Degreaser,32oz Bottle,RD
SDP
Catalog No. 5000790394
Encompass_Preferred
Low-residue, low-foaming, concentrated cleaner is specially formulated for daily maintenance of high-traffic surfaces or scrub/recoat applications. Also use it as a surface cleaner or degreaser. For best results, pair with SC Johnson Professional Hyper Concentrate Floor Stripper and Multi-Surface Floor Finish (none included). Use this Heavy Duty Neutral pH Floor & Surface Cleaner on a variety of flooring types such as vinyl composite, vinyl asphalt, vinyl asbestos, 100 percent sheet vinyl, terrazzo, rubber, linoleum, marble, granite, concrete and sealed wood floors. Squeeze-and-pour bottle includes a measuring cup for easy cleaning.
Catalog No. 50-007-90394 Supplier Enterprise Technology Solutions Supplier No. SJN680081
Low-residue, low-foaming, concentrated cleaner is specially formulated for daily maintenance of high-traffic surfaces or scrub/recoat applications. Also use it as a surface cleaner or degreaser. For best results, pair with SC Johnson Professional Hyper Concentrate Floor Stripper and Multi-Surface Floor Finish (none included). Use this Heavy Duty Neutral pH Floor & Surface Cleaner on a variety of flooring types such as vinyl composite, vinyl asphalt, vinyl asbestos, 100 percent sheet vinyl, terrazzo, rubber, linoleum, marble, granite, concrete and sealed wood floors. Squeeze-and-pour bottle includes a measuring cup for easy cleaning.