Learn More
Enterprise Technology Solutions S.C. JOHNSON & SON, INC Floor Cleaner,TruFill Cartridge,Concentrate,2-Liter,RD
SDP
Catalog No. 5000790395
Encompass_Preferred
TruFill Heavy Duty Neutral Floor Cleaner is a concentrated formula in a portable, no-spill, 2-liter cartridge for TruFill dispensing systems. Low-residue, low-foaming concentrated cleaner is formulated for daily maintenance of high-traffic floor surfaces or scrub/recoat applications. Integrated metering system provides worry-free dilution delivery for up to 271 in-use gallons. Use this cartridge with a TruFill dispenser head and TruFill hose (none included) on a variety of floor types such as vinyl composite, vinyl asphalt, vinyl asbestos, vinyl, terrazzo, rubber, linoleum, marble, granite, concrete and sealed wood floors.
Catalog No. 50-007-90395 Supplier Enterprise Technology Solutions Supplier No. SJN684502
TruFill Heavy Duty Neutral Floor Cleaner is a concentrated formula in a portable, no-spill, 2-liter cartridge for TruFill dispensing systems. Low-residue, low-foaming concentrated cleaner is formulated for daily maintenance of high-traffic floor surfaces or scrub/recoat applications. Integrated metering system provides worry-free dilution delivery for up to 271 in-use gallons. Use this cartridge with a TruFill dispenser head and TruFill hose (none included) on a variety of floor types such as vinyl composite, vinyl asphalt, vinyl asbestos, vinyl, terrazzo, rubber, linoleum, marble, granite, concrete and sealed wood floors.