Learn More
Enterprise Technology Solutions S.C. JOHNSON & SON, INC Hand Cleaner, Tube, 250 ml, Brown
SDP
Catalog No. 5000789997
Encompass_Preferred
Kresto Classic Hand Cleaner features a toxicologically safe, heavy-duty formula to effectively remove extremely heavy soiling from your hands. Specialist solvent offers excellent cleansing properties even with super-heavy contamination. Eucornol emulsifying active increases the performance of the product to remove contaminants from the skin and reduce the risk of skin irritation. Astopon natural scrubbing agent is produced from refined walnut shell powder to rinse easily away and prevent blocked drains and pipes.
Catalog No. 50-007-89997 Supplier Enterprise Technology Solutions Supplier No. SJNKCL250ML
Kresto Classic Hand Cleaner features a toxicologically safe, heavy-duty formula to effectively remove extremely heavy soiling from your hands. Specialist solvent offers excellent cleansing properties even with super-heavy contamination. Eucornol emulsifying active increases the performance of the product to remove contaminants from the skin and reduce the risk of skin irritation. Astopon natural scrubbing agent is produced from refined walnut shell powder to rinse easily away and prevent blocked drains and pipes.