Learn More
Enterprise Technology Solutions S.C. JOHNSON & SON, INC Hand Cleaner, Tube, 250 ml, Brown
SDP
Supplier:  Enterprise Technology Solutions SJNKSP250ML
Encompass_Preferred
Kresto Special Ultra Hand Cleaner features an ester-based solvent to remove very adherent substances while providing outstanding cleaning action. Highly concentrated formula allows you to use a small amount of cleaner for effective and economical cleaning. Astopon natural scrubbing agent is produced from refined walnut shell powder to rinse easily away and prevent blocked drains and pipes. Silicone-free soap will have no adverse effect if it comes in contact with surfaces before the application of paint or surface coatings. Effective specialist cleaning formula removes alkyd and acrylic paints, base coats, lacquers, resins, body fillers, adhesives and varnishes.
Catalog No. 50-007-90331