Learn More
Enterprise Technology Solutions S.C. JOHNSON & SON, INC Hand Sanitizer, Foam, Non-Alcohol, 1 Liter, 6/CT, Clear
SDP
Catalog No. 5000789963
Encompass_Preferred
Designed for use in SC Johnson 1 liter hand sanitizer dispensers, this foam hand sanitizer features a formula that is not alcohol-based so it is safe for use in spaces where flammable products are a concern. Sanitizing ingredients kill up to 99.99 percent of many common germs in 30 seconds without the use of soap and water. Skin conditioners prevent your skin from drying out to keep hands feeling smooth. Dye-free, perfume-free sanitizer helps reduce potential for allergic reaction and skin irritation. Instant foam action provides complete control of the product to prevent dripping and splashing. Clinically proven, skin-friendly sanitizer has been independently tested and proven to prevent skin dryness even after frequent use.
Catalog No. 50-007-89963 Supplier Enterprise Technology Solutions Supplier No. SJN55857
Designed for use in SC Johnson 1 liter hand sanitizer dispensers, this foam hand sanitizer features a formula that is not alcohol-based so it is safe for use in spaces where flammable products are a concern. Sanitizing ingredients kill up to 99.99 percent of many common germs in 30 seconds without the use of soap and water. Skin conditioners prevent your skin from drying out to keep hands feeling smooth. Dye-free, perfume-free sanitizer helps reduce potential for allergic reaction and skin irritation. Instant foam action provides complete control of the product to prevent dripping and splashing. Clinically proven, skin-friendly sanitizer has been independently tested and proven to prevent skin dryness even after frequent use.