Learn More
Enterprise Technology Solutions S.C. JOHNSON & SON, INC Hand Soap, Antibacterial, Foam, 1.2 Liter, 3/CT, Clear
SDP
Catalog No. 5000830054
Encompass_Preferred
Foam hand soap features an antibacterial formula to cleanse your skin and kill bacteria. Skin conditioners help your skin feel smooth and firm after each use. Triclosan-free formulation contains benzalkonium chloride as an active agent. Perfume-free, dye-free soap will not taint food and is suitable for all food-handling environments. Mild design is excellent for frequent use. Foam hand soap is perfect for removing light to medium contamination in low-risk and high-risk areas where high standards of hand hygiene are required.
Catalog No. 50-008-30054 Supplier Enterprise Technology Solutions Supplier No. SJNAGB120TF
Foam hand soap features an antibacterial formula to cleanse your skin and kill bacteria. Skin conditioners help your skin feel smooth and firm after each use. Triclosan-free formulation contains benzalkonium chloride as an active agent. Perfume-free, dye-free soap will not taint food and is suitable for all food-handling environments. Mild design is excellent for frequent use. Foam hand soap is perfect for removing light to medium contamination in low-risk and high-risk areas where high standards of hand hygiene are required.