Learn More
Enterprise Technology Solutions S.C. JOHNSON & SON, INC Hand Soap,f/Grease,Heavy-Duty,Scrub.,Solvent-Free,2L,6/CT,BG
SDP
Catalog No. 5000828740
Encompass_Preferred
Solopol Classic Heavy-Duty Hand Cleaner removes heavy oils, grease and water-based lacquers. Specially engineered, solvent-free formula provides an unexpectedly high-level cleaning performance. Optimally blended surfactants create a mixed-micellar cleaning system that penetrates and lifts heavy soiling. Unique cleaning blend also solubilizes a wide range of ingrained oil, grease, carbon black, lubricants and resins. Made from walnut shell powder, Astopon scrubbing agent easily rinses away to prevent clocked drains and pipes.
Catalog No. 50-008-28740 Supplier Enterprise Technology Solutions Supplier No. SJNPN98318706
Solopol Classic Heavy-Duty Hand Cleaner removes heavy oils, grease and water-based lacquers. Specially engineered, solvent-free formula provides an unexpectedly high-level cleaning performance. Optimally blended surfactants create a mixed-micellar cleaning system that penetrates and lifts heavy soiling. Unique cleaning blend also solubilizes a wide range of ingrained oil, grease, carbon black, lubricants and resins. Made from walnut shell powder, Astopon scrubbing agent easily rinses away to prevent clocked drains and pipes.