Learn More
Enterprise Technology Solutions S.C. JOHNSON & SON, INC Hand Soap,f/Grease/Oil,Classic,Mild Solvent,Scrub,2L,6/CT,BG
SDP
Catalog No. 5000830056
Encompass_Preferred
Super heavy-duty hand cleaner easily removes difficult, deeply ingrained oil contaminants. Specially engineered formula uses mild solvent Isohexadecane and Eucornol emulsifying active. These important ingredients in this solvent-based cleaner help create hand soap with strong cleaning power and low potential to cause skin irritation. Optimally blended surfactants create a mixed-micellar cleaning system that penetrates and lifts heavy soiling. Unique cleaning blend also solubilizes a wide range of ingrained oil, grease, carbon black, lubricants and resins. Made from walnut shell powder, Astopon scrubbing agent easily rinses away to prevent clogged drains and pipes. Low-odor fragrance doesn't linger for a more pleasant experience.
Catalog No. 50-008-30056 Supplier Enterprise Technology Solutions Supplier No. SJNPN98704506
Super heavy-duty hand cleaner easily removes difficult, deeply ingrained oil contaminants. Specially engineered formula uses mild solvent Isohexadecane and Eucornol emulsifying active. These important ingredients in this solvent-based cleaner help create hand soap with strong cleaning power and low potential to cause skin irritation. Optimally blended surfactants create a mixed-micellar cleaning system that penetrates and lifts heavy soiling. Unique cleaning blend also solubilizes a wide range of ingrained oil, grease, carbon black, lubricants and resins. Made from walnut shell powder, Astopon scrubbing agent easily rinses away to prevent clogged drains and pipes. Low-odor fragrance doesn't linger for a more pleasant experience.