Learn More
Enterprise Technology Solutions S.C. JOHNSON & SON, INC Soap, Hand Hair Body, 1 Liter, 6/CT, Blue
SDP
Catalog No. 5000829158
Encompass_Preferred
Estesol Hand, Hair and Body Cleaner features a three-in-one formula to remove light duty oils, greases, general grime and soilings from your hands, hair and body. Rainforest fragrance provides a refreshing, invigorating scent that's appealing to both males and females. Mild yet effective blend of cleansing agents are suitable for frequent use on hair and body. Low pH value helps minimize disruption from use to the skin's acid mantle. Mild preservatives reduce the risk of skin irritation. Silicone-free soap will have no adverse effect if it comes in contact with surfaces before the application of paint or surface coatings. Added skin moisturizer helps prevent drying and leaves the skin feeling smooth after use. Hand, hair and body cleaner is designed for SC Johnson Cleanse Washroom Dispensers and Proline Curve Manual Dispensers (each sold separately).
Catalog No. 50-008-29158 Supplier Enterprise Technology Solutions Supplier No. SJNHAB1L
Estesol Hand, Hair and Body Cleaner features a three-in-one formula to remove light duty oils, greases, general grime and soilings from your hands, hair and body. Rainforest fragrance provides a refreshing, invigorating scent that's appealing to both males and females. Mild yet effective blend of cleansing agents are suitable for frequent use on hair and body. Low pH value helps minimize disruption from use to the skin's acid mantle. Mild preservatives reduce the risk of skin irritation. Silicone-free soap will have no adverse effect if it comes in contact with surfaces before the application of paint or surface coatings. Added skin moisturizer helps prevent drying and leaves the skin feeling smooth after use. Hand, hair and body cleaner is designed for SC Johnson Cleanse Washroom Dispensers and Proline Curve Manual Dispensers (each sold separately).