Learn More
Enterprise Technology Solutions S.C. JOHNSON & SON, INC UV Skin Protection Cream, Tube, 100 ml, White
SDP
Catalog No. 5000789879
Encompass_Preferred
UV skin-protection cream features a high sun protection factor of 30 to provide 30 times the skin's normal protection against the sun's UVB rays. UVC protection guards against UVC rays created artificially during some industrial processes. Skin-protection cream has passed lineman's glove testing and is safe to be used by outdoor electricians wearing electrical insulating gloves. Water-resistant formula withstands heavy perspiration or immersion in water. Cream quickly and easily absorbs to encourage regular use and does not affect dexterity with handheld tools. Perfume-free formula helps reduce potential for allergic reaction and skin irritation.
Catalog No. 50-007-89879 Supplier Enterprise Technology Solutions Supplier No. SJNSUN100ML
UV skin-protection cream features a high sun protection factor of 30 to provide 30 times the skin's normal protection against the sun's UVB rays. UVC protection guards against UVC rays created artificially during some industrial processes. Skin-protection cream has passed lineman's glove testing and is safe to be used by outdoor electricians wearing electrical insulating gloves. Water-resistant formula withstands heavy perspiration or immersion in water. Cream quickly and easily absorbs to encourage regular use and does not affect dexterity with handheld tools. Perfume-free formula helps reduce potential for allergic reaction and skin irritation.