Learn More
Bellco Glass, Inc. S/Cap Cult Tube Complete, 13x100mm Bulk-Pack - Bellco logo
SDP
Catalog No. 501945294
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Includes long skirted black phenolic screw cap with white rubber liner. MADE IN THE USA. Screw cap included Size: 13 x 100mm
Catalog No. 50-194-5294 Supplier Bellco Glass, Inc. Supplier No. 201213000
Manufactured from type 1 class a Borosilicate glass. Includes long skirted black phenolic screw cap with white rubber liner. MADE IN THE USA. Screw cap included Size: 13 x 100mm