Learn More
Bellco Glass, Inc. S/Cap Grad Erlenmeyer Flask ,1000mL With 38mm Cap
SDP
Catalog No. 501945478
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Ideal for culture aerobic & anaerobic organisms, media storage and mixing. Deep skirted screw caps protect against contamination. Supplied with rubber line black phenolic caps and graduated to approximate volumes.. MADE IN THE USA Size: 1L Cap: 38-415 Dimensions: 128x220mm
Catalog No. 50-194-5478 Supplier Bellco Glass, Inc. Supplier No. 251401000
Manufactured from type 1 class a Borosilicate glass. Ideal for culture aerobic & anaerobic organisms, media storage and mixing. Deep skirted screw caps protect against contamination. Supplied with rubber line black phenolic caps and graduated to approximate volumes.. MADE IN THE USA Size: 1L Cap: 38-415 Dimensions: 128x220mm