Learn More
Bellco Glass, Inc. S/Cap Grad Erlenmeyer Flask ,125mL With 25mm Cap
SDP
Catalog No. 501945474
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Ideal for culture aerobic & anaerobic organisms, media storage and mixing. Deep skirted screw caps protect against contamination. Supplied with rubber line black phenolic caps and graduated to approximate volumes.. MADE IN THE USA Size: 50mL Cap: 24-415 Dimensions: 50x85mm
Catalog No. 50-194-5474 Supplier Bellco Glass, Inc. Supplier No. 251400125
Manufactured from type 1 class a Borosilicate glass. Ideal for culture aerobic & anaerobic organisms, media storage and mixing. Deep skirted screw caps protect against contamination. Supplied with rubber line black phenolic caps and graduated to approximate volumes.. MADE IN THE USA Size: 50mL Cap: 24-415 Dimensions: 50x85mm