Learn More
Bellco Glass, Inc. S/Cap Grad Erlenmeyer Flask ,25mL 18mm Screw Cap
SDP
Catalog No. 501945472
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Ideal for culture aerobic & anaerobic organisms, media storage and mixing. Deep skirted screw caps protect against contamination. Supplied with rubber line black phenolic caps and graduated to approximate volumes. . MADE IN THE USA Size: 25mL Cap: 18-415 Dimensions: 41x65mm
Catalog No. 50-194-5472 Supplier Bellco Glass, Inc. Supplier No. 251400025
Manufactured from type 1 class a Borosilicate glass. Ideal for culture aerobic & anaerobic organisms, media storage and mixing. Deep skirted screw caps protect against contamination. Supplied with rubber line black phenolic caps and graduated to approximate volumes. . MADE IN THE USA Size: 25mL Cap: 18-415 Dimensions: 41x65mm