Learn More
Bellco Glass, Inc. S/Cap Grad Erlenmeyer Flask ,3000mL With 38mm Cap
SDP
Catalog No. 501945480
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Ideal for culture aerobic & anaerobic organisms, media storage and mixing. Deep skirted screw caps protect against contamination. Supplied with rubber line black phenolic caps and graduated to approximate volumes. . MADE IN THE USA Size: 3L Cap: 38-415 Dimensions: 184x320mm
Catalog No. 50-194-5480 Supplier Bellco Glass, Inc. Supplier No. 251403000
Manufactured from type 1 class a Borosilicate glass. Ideal for culture aerobic & anaerobic organisms, media storage and mixing. Deep skirted screw caps protect against contamination. Supplied with rubber line black phenolic caps and graduated to approximate volumes. . MADE IN THE USA Size: 3L Cap: 38-415 Dimensions: 184x320mm