Learn More
Bellco Glass, Inc. S/Cap Grad Erlenmeyer Flask ,300mL With 38mm Cap
SDP
Catalog No. 501945476
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Ideal for culture aerobic & anaerobic organisms, media storage and mixing. Deep skirted screw caps protect against contamination. Supplied with rubber line black phenolic caps and graduated to approximate volumes.. MADE IN THE USA Size: 300mL Cap: 38-415 Dimensions: 88x150mm
Catalog No. 50-194-5476 Supplier Bellco Glass, Inc. Supplier No. 251400300
Manufactured from type 1 class a Borosilicate glass. Ideal for culture aerobic & anaerobic organisms, media storage and mixing. Deep skirted screw caps protect against contamination. Supplied with rubber line black phenolic caps and graduated to approximate volumes.. MADE IN THE USA Size: 300mL Cap: 38-415 Dimensions: 88x150mm