Learn More
Bellco Glass, Inc. S/Cap Grad Erlenmeyer Flask ,50mL With 25mm Screw Cap
SDP
Catalog No. 501945473
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Ideal for culture aerobic & anaerobic organisms, media storage and mixing. Deep skirted screw caps protect against contamination. Supplied with rubber line black phenolic caps and graduated to approximate volumes. . MADE IN THE USA Size: 25mL Cap: 18-415 Dimensions: 41x65mm
Catalog No. 50-194-5473 Supplier Bellco Glass, Inc. Supplier No. 251400050
Manufactured from type 1 class a Borosilicate glass. Ideal for culture aerobic & anaerobic organisms, media storage and mixing. Deep skirted screw caps protect against contamination. Supplied with rubber line black phenolic caps and graduated to approximate volumes. . MADE IN THE USA Size: 25mL Cap: 18-415 Dimensions: 41x65mm