Learn More
Bellco Glass, Inc. S/Cap Grad Erlenmeyer Flask ,6000mL With 38mm Cap
SDP
Supplier:  Bellco Glass, Inc. 251406000
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Ideal for culture aerobic & anaerobic organisms, media storage and mixing. Deep skirted screw caps protect against contamination. Supplied with rubber line black phenolic caps and graduated to approximate volumes.. MADE IN THE USA Size: 6L Cap: 38-415 Dimensions: 235x390mm
Catalog No. 50-194-5482