Learn More
Bellco Glass, Inc. Screw Cap Leighton Tube,19X105mm
SDP
Catalog No. 501944676
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Includes long skirted screw cap with nontoxic white rubber liner. Extra long working area yet short overall length for convenient handling. The short neck provides easy entry and removal of cover slips. MADE IN THE USA
Catalog No. 50-194-4676 Supplier Bellco Glass, Inc. Supplier No. 190419105
Manufactured from type 1 class a Borosilicate glass. Includes long skirted screw cap with nontoxic white rubber liner. Extra long working area yet short overall length for convenient handling. The short neck provides easy entry and removal of cover slips. MADE IN THE USA