Learn More
Bellco Glass, Inc. S/Cap Roux Cult Bottle,1200mL Schott Blank
SDP
Catalog No. 501945742
Encompass_Preferred
Ideal for large area mono-layer cell culture growth. Offset neck prevents culture from entering neck area. Working volume is approximately one half total volume. Neck is designed for use with 38mm deep skirted non-toxic rubber lined black phenolic screw cap included. ~ DIMENSIONS: 132mm OD X 56mm W X 280mm OAH 2 per case
Catalog No. 50-194-5742 Supplier Bellco Glass, Inc. Supplier No. 563212000
Ideal for large area mono-layer cell culture growth. Offset neck prevents culture from entering neck area. Working volume is approximately one half total volume. Neck is designed for use with 38mm deep skirted non-toxic rubber lined black phenolic screw cap included. ~ DIMENSIONS: 132mm OD X 56mm W X 280mm OAH 2 per case