Learn More
Medchemexpress LLC HY-B0006B 5mg Medchemexpress, (S)-Carvedilol CAS:95094-00-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0006B5MG
Encompass_Preferred
Medchemexpress, HY-B0006B 5mg (S)-Carvedilol CAS:95094-00-1 (S)-Carvedilol, the S-enantiomer of Carvedilol, is a non-selective β/α-1 blocker. (S)-Carvedilol exerts protection against the vascular or cardiac toxicity of Doxorubicin (DOX). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2342