Learn More
Medchemexpress LLC (S)-carvedilol | 95094-00-1 | 99.6% | 406.47 | C24H26N2O4 | 10 MG
SDP
Catalog No. 5000298411
Encompass_Preferred
(S)-Carvedilol is the S-enantiomer of carvedilol, a non-selective beta and alpha-1 adrenergic receptor blocker used in research to evaluate cardiovascular and vascular pharmacology. It has been reported to protect against doxorubicin-induced vascular and cardiac toxicity and is supplied at high purity for analytical and pharmacological studies.
  • Non-selective beta and alpha-1 adrenergic blocker.
  • Stereochemically defined S-enantiomer for enantioselective studies.
  • High purity suitable for research use.
  • Available as solid quantities and as a DMSO solution for assays.
  • Used in studies of cardioprotection and vascular toxicity mitigation.
Catalog No. 50-002-98411 Supplier Medchemexpress LLC Supplier No. HYB0006B10MG
(S)-Carvedilol is the S-enantiomer of carvedilol, a non-selective beta and alpha-1 adrenergic receptor blocker used in research to evaluate cardiovascular and vascular pharmacology. It has been reported to protect against doxorubicin-induced vascular and cardiac toxicity and is supplied at high purity for analytical and pharmacological studies.
  • Non-selective beta and alpha-1 adrenergic blocker.
  • Stereochemically defined S-enantiomer for enantioselective studies.
  • High purity suitable for research use.
  • Available as solid quantities and as a DMSO solution for assays.
  • Used in studies of cardioprotection and vascular toxicity mitigation.