Learn More
Apexbio Technology LLC S-CARVONE-1ML
SDP
Catalog No. 5000569244
Encompass_Preferred
(S)-( )-Carvone (CAS No 2244-16-8) is a naturally occurring monoterpene commonly studied in the field of biomedical research It is derived from essential oils of caraway seeds and spearmint leaves Its mechanism of action involves interaction with GABA receptors and modulation of ion channel pathways contributing to its potential as an anticonvulsant agent In vitro studies often demonstrate activity in the nanomolar to low micromolar range This compound is frequently used in diverse applications such as evaluating its effects on neuronal cells and exploring its anti-inflammatory and antimicrobial properties in various cell models Additionally it is examined in animal studies for its potential therapeutic effects and included in drug screening protocols to assess its efficacy and safety profiles Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-69244 Supplier Apexbio Technology LLC Supplier No. C71371
(S)-( )-Carvone (CAS No 2244-16-8) is a naturally occurring monoterpene commonly studied in the field of biomedical research It is derived from essential oils of caraway seeds and spearmint leaves Its mechanism of action involves interaction with GABA receptors and modulation of ion channel pathways contributing to its potential as an anticonvulsant agent In vitro studies often demonstrate activity in the nanomolar to low micromolar range This compound is frequently used in diverse applications such as evaluating its effects on neuronal cells and exploring its anti-inflammatory and antimicrobial properties in various cell models Additionally it is examined in animal studies for its potential therapeutic effects and included in drug screening protocols to assess its efficacy and safety profiles Experimental concentrations typically depend on the specific research design