Learn More
Medchemexpress LLC (S)-(+)-Carvone | 2244-16-8 | 99.3% | 150.22 | 1 G
SDP
Catalog No. 5000454495
Encompass_Preferred
(S)-(+)-Carvone is an orally active natural product used for research. It increases antioxidant enzyme (SOD, CAT) activity and ROS, reducing oxidative stress markers (MDA, AChE). It also lowers proinflammatory cytokines (IL-1β, IL-4, IL-6, IL-10), downregulates NLRP3, and induces apoptotic death by increasing caspase-8, -9, and -3 activities. It demonstrates antimanic-like effects, liver protection, and anticancer activity against skin cancer, also improving memory and arthritis.
  • Increases antioxidant enzyme (SOD, CAT) activity and ROS
  • Reduces oxidative stress markers (MDA, AChE)
  • Lowers proinflammatory cytokines (IL-1β, IL-4, IL-6, IL-10) and downregulates NLRP3
  • Induces apoptotic death by increasing caspase-8, -9, and -3 activities
  • Demonstrates antimanic-like effects, liver protection, and anticancer activity against skin cancer
  • Improves memory and arthritis
Catalog No. 50-004-54495 Supplier Medchemexpress LLC Supplier No. HYW0135791G
(S)-(+)-Carvone is an orally active natural product used for research. It increases antioxidant enzyme (SOD, CAT) activity and ROS, reducing oxidative stress markers (MDA, AChE). It also lowers proinflammatory cytokines (IL-1β, IL-4, IL-6, IL-10), downregulates NLRP3, and induces apoptotic death by increasing caspase-8, -9, and -3 activities. It demonstrates antimanic-like effects, liver protection, and anticancer activity against skin cancer, also improving memory and arthritis.
  • Increases antioxidant enzyme (SOD, CAT) activity and ROS
  • Reduces oxidative stress markers (MDA, AChE)
  • Lowers proinflammatory cytokines (IL-1β, IL-4, IL-6, IL-10) and downregulates NLRP3
  • Induces apoptotic death by increasing caspase-8, -9, and -3 activities
  • Demonstrates antimanic-like effects, liver protection, and anticancer activity against skin cancer
  • Improves memory and arthritis